Learn More
Palosuran, MedChemExpress
Product Code. 16485056
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485056 100 mg 100mg
16474088 10 mg 10mg
16450447 50 mg 50mg
16416797 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16485056 Supplier Medchem Express Supplier No. HY10655
MedChemExpress Palosuran (ACT-058362) is a potent, selective, and orally active antagonist of urotensin II receptor, with an IC50 of 3.6 nM for CHO cell membranes expressing human recombinant receptors. Palosuran can improves pancreatic and renal function in diabetic rats.
Palosuran (ACT-058362) is a potent, selective, and orally active antagonist of urotensin II receptor, with an IC50 of 3.6 nM for CHO cell membranes expressing human recombinant receptors. Palosuran can improves pancreatic and renal function in diabetic rats.
Specifications
Chemical Name or Material Palosuran
CAS 540769-28-6
Color White
Purity Grade Notes Research
Molecular Formula C25H30N4O2
Quantity 100 mg
Synonym ACT-058362
Solubility Information DMSO : 50 mg/mL (119.47 mM; Need ultrasonic)
SMILES OC(CC1=CC=CC=C1)(CC2)CCN2CCNC(NC3=CC(C)=NC4=CC=CC=C34)=O
Molecular Weight (g/mol) 418.53
Formula Weight 418.53
Percent Purity 99.01%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less
Research purposes only