missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALS1/MPP5 Antibody (OTI2D3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73234AF350
This item is not returnable.
View return policy
Description
PALS1/MPP5 Monoclonal specifically detects PALS1/MPP5 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| PALS1/MPP5 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MPP5 | |
| Full length recombinant protein of human PALS1/MPP5 (NP_071919) produced in HEK293T cells. | |
| 0.1 mL | |
| 64398.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI2D3 | |
| Western Blot, Flow Cytometry | |
| MAGUK p55 subfamily member 5, membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5), PALS1FLJ12615, stardust | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction