missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALS1/MPP5 Antibody (OTI2D3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73234
This item is not returnable.
View return policy
Description
PALS1/MPP5 Monoclonal specifically detects PALS1/MPP5 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| PALS1/MPP5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, CyTOF-ready | |
| MAGUK p55 subfamily member 5, membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5), PALS1FLJ12615, stardust | |
| Mouse | |
| 77.1 kDa | |
| 100 μg | |
| 64398 | |
| Human, Mouse, Rat, Canine | |
| Lyophilized |
| Western Blot, Flow Cytometry, CyTOF | |
| OTI2D3 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| MPP5 | |
| Full length human recombinant protein of human MPP5(NP_071919) produced in HEK293T cell. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction