missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan Actin Antibody (5J11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25142F
This item is not returnable.
View return policy
Description
pan Actin Monoclonal specifically detects pan Actin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| pan Actin | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| ACTA1 | |
| An Actin preparation derived from bovine brain. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
| 58 | |
| Human, Mouse, Rat, Pig, Bovine, Equine | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 5J11 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody recognizes all six isotypes of Actin. | |
| Store at 4C in the dark. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto