missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan Cadherin Antibody (OTI4F1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73237JF646
This item is not returnable.
View return policy
Description
pan Cadherin Monoclonal specifically detects pan Cadherin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| pan Cadherin | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CDH1 | |
| Full length recombinant protein of human pan Cadherin (NP_004351) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4F1 | |
| Western Blot, Immunohistochemistry | |
| Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| 999.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction