missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan Endothelial Cell Marker Antibody (RM0061-4J13), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12438AF750
This item is not returnable.
View return policy
Description
Pan Endothelial Cell Marker Monoclonal specifically detects Pan Endothelial Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Pan Endothelial Cell Marker | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Cell lysates of mouse endothelial cells | |
| 0.1 mL | |
| Mouse | |
| IgG2 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0061-4J13 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction