missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan Endothelial Cell Marker Antibody (RM0061-4J13), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12438V
This item is not returnable.
View return policy
Description
Pan Endothelial Cell Marker Monoclonal specifically detects Pan Endothelial Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Pan Endothelial Cell Marker | |
| Monoclonal | |
| DyLight 405 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0061-4J13 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Cell lysates of mouse endothelial cells | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction