Learn More
Pan-Endothelium Marker, p Ser124, p Ser123 Antibody (HIS52), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-00076AF647
Product Code. 18060977
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Pan-Endothelium Marker Monoclonal specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pan-Endothelium Marker (p Ser123, p Ser124, p ser123, p ser124)
Monoclonal
Alexa Fluor 647
Mouse
Affinity Purified
RUO
Rat
IgG1 κ
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
HIS52
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative)
The immunogen for this antibody is Pan endothelium.
0.1 mL
Primary
Store at 4C in the dark.