Learn More
Pan-Endothelium Marker, p Ser124, p Ser123 Antibody (HIS52), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00076AF647
This item is not returnable.
View return policy
Description
Pan-Endothelium Marker Monoclonal specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative).
Specifications
| Pan-Endothelium Marker (p Ser123, p Ser124, p ser123, p ser124) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Rat | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| HIS52 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative) | |
| The immunogen for this antibody is Pan endothelium. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |