missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan-Endothelium Marker, p Ser124, p Ser123 Antibody (HIS52), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00076JF646
This item is not returnable.
View return policy
Description
Pan-Endothelium Marker Monoclonal specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Pan-EndotheliumMarker | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| The immunogen for this antibody is Pan endothelium. | |
| 0.1 mL | |
| Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| HIS52 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction