missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Pancreas Tissue Slides (Adult Normal)- Frozen
Shop All Bio Techne Products
Click to view available options
Quantity:
5 Slides
Unit Size:
Pack of 5
Specifications
Specifications
| Quantity | 5 Slides |
| For Use With (Application) | Immunohistochemistry (IHC), Immunohistochemistry (Frozen), ISH |
| Species | Human |
| Target Specificity | Pancreas |
| Content And Storage | Store at -70°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction