missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Amylase Alpha Antibody (OTI6D4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73238CL1
This item is not returnable.
View return policy
Description
Pancreatic Amylase Alpha Monoclonal antibody specifically detects Pancreatic Amylase Alpha in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Pancreatic Amylase Alpha | |
| Monoclonal | |
| CoraFluor 1 | |
| AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase | |
| Recombinant protein expressed in E.coli corresponding to amino acids 300-511 of human AMY2A | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI6D4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 279 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction