missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Amylase Beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19687
This item is not returnable.
View return policy
Description
Pancreatic Amylase Beta Polyclonal specifically detects Pancreatic Amylase Beta in Human samples. It is validated for Western Blot.
Specifications
| Pancreatic Amylase Beta | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| alpha-amylase 2B, AMY2, AMY2B amylase, alpha 2B (pancreatic) | |
| Rabbit | |
| 58 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| P19961 | |
| AMY2B | |
| Recombinant protein encompassing a sequence within the center region of human alpha amylase 2B (pancreatic). The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 280 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction