missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Lipase Antibody (120), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90231AF488
This item is not returnable.
View return policy
Description
Pancreatic Lipase Monoclonal specifically detects Pancreatic Lipase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Pancreatic Lipase | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5406 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 120 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Pancreatic Lipase (P16233; Met1-Cys465). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction