missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Lipase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00153-100ul
This item is not returnable.
View return policy
Description
Pancreatic Lipase Polyclonal antibody specifically detects Pancreatic Lipase in Human samples. It is validated for ELISA
Specifications
| Pancreatic Lipase | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase | |
| Produced in rabbits immunized with purified, recombinant Human Pancreatic Lipase (Uniprot#: P16233; Met1-Cys465) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5406 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction