missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Pancreatic Lipase Antibody (PNLIP/9039), Novus Biologicals™

Mouse Monoclonal Antibody

€ 232.00 - € 462.00

Specifications

Antigen Pancreatic Lipase
Clone PNLIP/9039
Concentration 0.2 mg/mL
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043085
View Documents
Novus Biologicals™
NBP3-26713-20ug
20 μg
€ 232.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043180
View Documents
Novus Biologicals™
NBP3-26713-100ug
100 μg
€ 462.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pancreatic Lipase Monoclonal antibody specifically detects Pancreatic Lipase in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications

Specifications

Pancreatic Lipase
0.2 mg/mL
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase
Recombinant fragment (around aa 1-200) of human Pancreatic Lipase protein (exact sequence is proprietary)
Primary
Store at 4°C.
PNLIP/9039
ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
5406
IgG2b κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.