missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Pancreatic Lipase Antibody (PNLIP/9040) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26989B
This item is not returnable.
View return policy
Description
Pancreatic Lipase Monoclonal antibody specifically detects Pancreatic Lipase in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| Pancreatic Lipase | |
| Monoclonal | |
| Biotin | |
| EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase | |
| Recombinant fragment (around aa 265-465) of human Pancreatic Lipase protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| PNLIP/9040 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5406 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction