missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Lipase Related Protein 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93552-0.02ml
This item is not returnable.
View return policy
Description
Pancreatic Lipase Related Protein 2 Polyclonal antibody specifically detects Pancreatic Lipase Related Protein 2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Pancreatic Lipase Related Protein 2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| EC 3.1.1, EC 3.1.1.3, Galactolipase, pancreatic lipase-related protein 2, PL-RP2, PLRP2EC 3.1.1.26 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 301-470 of human PNLIPRP2 (NP_005387.2). LGYPCASYDEFQESKCFPCPAEGCPKMGHYADQFKGKTSAVEQTFFLNTGESGNFTSWRYKVSVTLSGKEKVNGYIRIALYGSNENSKQYEIFKGSLKPDASHTCAIDVDFNVGKIQKVKFLWNKRGINLSEPKLGASQITVQSGEDGTEYNFCSSDTVEENVLQSLYPC | |
| 0.02 mL | |
| Cancer | |
| 5408 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction