missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Polypeptide (PPY) Monoclonal Antibody (OTI1G2), OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813060S
This item is not returnable.
View return policy
Description
Pancreatic Polypeptide (PPY) Monoclonal antibody specifically detects Pancreatic Polypeptide (PPY) in Human samples. It is validated for Western BlotSpecifications
| Pancreatic Polypeptide (PPY) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01298 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5539 | |
| -20°C | |
| IgG2a |
| Western Blot | |
| OTI1G2 | |
| Unconjugated | |
| PPY | |
| AV061413; C-terminal peptide; HGNC:9327; IGSPP; IOPPP; Obinepitide; panceatic polypeptide; Pancreatic hormone; Pancreatic icosapeptide; Pancreatic polypeptide; pancreatic polypeptide Y; pancreatic prohormone; Pancreatic prohormone percursor; pancreatic prohormone precursor; PH; PI; PNP; PP; PP1; PPA1; PPase; Ppy; prepro-PP (prepropancreatic polypeptide); SID6-8061 | |
| Human recombinant protein fragment corresponding to amino acids 30-88 of human PPY (NP_002713) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction