missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic trypsin inhibitor Antibody (21), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23589AF532
This item is not returnable.
View return policy
Description
Pancreatic trypsin inhibitor Monoclonal specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA.
Specifications
| Pancreatic trypsin inhibitor | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PTI | |
| Aprotinin from bovine lung adsorbed onto aluminum hydroxide gel | |
| Primary | |
| Clone 21 binds bovine aprotinin immobilized on a solid phase or free in solution. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA | |
| 21 | |
| Western Blot, ELISA | |
| Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor | |
| Mouse | |
| 0.1 mL | |
| 404172 | |
| Bovine | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction