missing translation for 'onlineSavingsMsg'
Learn More

Pancreatin U.S.P., MP Biomedicals™

Catalog Number p-4404470
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
8049-47-6
Diastase vera
This item is not returnable. View return policy
8049-47-6
Diastase vera
Color Tan to White
Quantity 100 g
Physical Form Powder
Chemical Name or Material Pancreatin
compliance-icons compliance-icons
Product Identifier
  • pancreatin USP
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : pancreatin USP
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'