missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Panendothelial Cell Antigen Antibody (MECA-32), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-77668DL594
This item is not returnable.
View return policy
Description
Panendothelial Cell Antigen Monoclonal antibody specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Panendothelial Cell Antigen | |
| Monoclonal | |
| DyLight 594 | |
| FELS, Fenestrated endothelial-linked structure protein, gp68, MECA32, MECA-32, Panendothelial Cell Antigen, plasmalemma vesicle associated protein, Plasmalemma Vesicle Protein 1, PV-1 protein | |
| Mouse lymph node stromal cells. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 84094 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MECA-32 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction