missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Panendothelial Cell Antigen Antibody (MECA-32), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-77668C
This item is not returnable.
View return policy
Description
Panendothelial Cell Antigen Monoclonal specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Panendothelial Cell Antigen | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Plvap | |
| Mouse lymph node stromal cells. | |
| 0.1 mL | |
| 84094 | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MECA-32 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| MECA32, MECA-32, Panendothelial Cell Antigen | |
| Rat | |
| Protein G purified | |
| Primary | |
| Pan endothelial cells except for CNS & muscle capillaries. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction