missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PANK2 Antibody (OTI3H9), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73241APC
This item is not returnable.
View return policy
Description
PANK2 Monoclonal specifically detects PANK2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| PANK2 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| PANK2 | |
| Full length recombinant protein of human PANK2 (NP_705902) produced in HEK293T cells. | |
| 0.1 mL | |
| 80025.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI3H9 | |
| Flow Cytometry | |
| C20orf48, EC 2.7.1.33, FLJ17232, Hallervorden-Spatz syndrome, HARPFLJ11729, hPanK2, HSS, MGC15053, NBIA1, pantothenate kinase 2, pantothenate kinase 2, mitochondrial, Pantothenic acid kinase 2, PKANneurodegeneration with brain iron accumulation 1 (Hallervorden-Spatz syndrome) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction