missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PANK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103155
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PANK2.
Pantothenate kinase is an essential regulatory enzyme in CoA biosynthesis, catalyzing the cytosolic phosphorylation of pantothenate (vitamin B5), N-pantothenoylcysteine, and pantetheine. CoA is the major acyl carrier, playing a central role in intermediary and fatty acid metabolism. In both yeast and fly, each with only 1 pantothenate kinase gene, the null mutant is inviable. Mutations in PANK2 are the cause of pantothenate kinase-associated neurodegeneration (PKAN), formerly known as Hallervorden-Spatz syndrome (HSS). PKAN is an autosomal recessive neurodegenerative disorder associated with iron accumulation in the brain. Mutations in PANK2 are the cause of hypoprebetalipoproteinemia, acanthocytosis, retinitis pigmentosa, and pallidal degeneration (HARP).
Specifications
| PANK2 | |
| Polyclonal | |
| Unconjugated | |
| PANK2 | |
| 4933409I19Rik; AI642621; C20orf48; Hallervorden-Spatz syndrome; HARP; hPanK2; HSS; NBIA1; PANK2; pantothenate kinase 2; pantothenate kinase 2 (Hallervorden-Spatz syndrome); pantothenate kinase 2, mitochondrial; pantothenate kinase 2, mitochondrial isoform 1 preproprotein; pantothenate kinase 2, mitochondrial-like; pantothenic acid kinase 2; PKAN | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80025 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BZ23 | |
| PANK2 | |
| A synthesized peptide derived from human PANK2(Accession Q9BZ23), corresponding to amino acid residues P16-R66. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction