missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PANK2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5114220

Product Code. 17192074

  • € 506.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ERFGLPGWAV ASSFGNMMSK EKREAVSKED LARATLITIT NNIGSIARMC For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 100%; Sheep: 100%; Zebrafish: 86%.

Pantothenate kinase is an essential regulatory enzyme in CoA biosynthesis, catalyzing the cytosolic phosphorylation of pantothenate (vitamin B5), N-pantothenoylcysteine, and pantetheine. CoA is the major acyl carrier, playing a central role in intermediary and fatty acid metabolism. In both yeast and fly, each with only 1 pantothenate kinase gene, the null mutant is inviable. Mutations in PANK2 are the cause of pantothenate kinase-associated neurodegeneration (PKAN), formerly known as Hallervorden-Spatz syndrome (HSS). PKAN is an autosomal recessive neurodegenerative disorder associated with iron accumulation in the brain. Mutations in PANK2 are the cause of hypoprebetalipoproteinemia, acanthocytosis, retinitis pigmentosa, and pallidal degeneration (HARP).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PANK2
Polyclonal
Unconjugated
PANK2
4933409I19Rik; AI642621; C20orf48; Hallervorden-Spatz syndrome; HARP; hPanK2; HSS; NBIA1; PANK2; pantothenate kinase 2; pantothenate kinase 2 (Hallervorden-Spatz syndrome); pantothenate kinase 2, mitochondrial; pantothenate kinase 2, mitochondrial isoform 1 preproprotein; pantothenate kinase 2, mitochondrial-like; pantothenic acid kinase 2; PKAN
Rabbit
Affinity chromatography
RUO
80025
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9BZ23
PANK2
Immunogen sequence: ERFGLPGWAV ASSFGNMMSK EKREAVSKED LARATLITIT NNIGSIARMC.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.