missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PANK3 Antibody (3H4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079646-M05
This item is not returnable.
View return policy
Description
PANK3 Monoclonal antibody specifically detects PANK3 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| PANK3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DKFZp686O22247, EC 2.7.1.33, FLJ12899, hPanK3, MGC16863, pantothenate kinase 3, Pantothenic acid kinase 3 | |
| PANK3 (NP_078870.1, 88 a.a. ∽ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PTQDLPTFIQMGRDKNFSTLQTVLCATGGGAYKFEKDFRTIGNLHLHKLDELDCLVKGLLYIDSVSFNGQAECYYFANASEPERCQKMPFNLD | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 3H4 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_078870 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 79646 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction