missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PANK3 Antibody (OTI3C4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73244
This item is not returnable.
View return policy
Description
PANK3 Monoclonal specifically detects PANK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| PANK3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500, Flow Cytometry 1:100, CyTOF-ready | |
| DKFZp686O22247, EC 2.7.1.33, FLJ12899, hPanK3, MGC16863, pantothenate kinase 3, Pantothenic acid kinase 3 | |
| Mouse | |
| 40.9 kDa | |
| 100 μg | |
| 79646 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot, Flow Cytometry, CyTOF | |
| OTI3C4 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| PANK3 | |
| Full length human recombinant protein of human PANK3 (NP_078870) produced in HEK293T cell. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction