missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PANK3 Antibody (OTI3C4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73244FR
This item is not returnable.
View return policy
Description
PANK3 Monoclonal specifically detects PANK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| PANK3 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| PANK3 | |
| Full length recombinant protein of human PANK3 (NP_078870) produced in HEK293T cells. | |
| 0.1 mL | |
| 79646.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI3C4 | |
| Western Blot, Flow Cytometry | |
| DKFZp686O22247, EC 2.7.1.33, FLJ12899, hPanK3, MGC16863, pantothenate kinase 3, Pantothenic acid kinase 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction