missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pannexin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA578455

Product Code. 16327154

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: U2OS Predicted Reactivity: Rat (81%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The pannexin gene family encodes a second class of putative gap junction proteins and are highly conserved in invertebrates and mammals. Pannexins (Panx) are four-pass transmembrane proteins that oligomerize to form large pore ion and metabolite-permeable channels. Pannexin-1 (PANX1) and Pannexin-3 are closely related, while Pannexin-2 is a more distant relation. PANX1 is a transmembrane protein that forms a mechanosensitive ATP-permeable channel between adjacent cells and in the endoplasmic reticulum. PANX1 may play a role as a Ca2+ -leak channel to regulate ER Ca2+ homeostasis and regulates neural stem and progenitor cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pannexin 1
Polyclonal
Unconjugated
PANX1
AI847747; innexin; MRS1; Pannexin 1; pannexin-1; Panx1; px1; UNQ2529; UNQ2529/PRO6028
Rabbit
Antigen affinity chromatography
RUO
24145
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.26 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Q96RD7
PANX1
Recombinant protein encompassing a sequence within the Intracellular domain of human Pannexin 1. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.