missing translation for 'onlineSavingsMsg'
Learn More

PAP-1, MedChemExpress

Product Code. 16435547
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435547

Brand: Medchem Express HY10015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PAP-1 (5-(4-Phenoxybutoxy)psoralen) is a potent, selective, and orally active Kv1.3 blocker (EC50=2 nM). PAP-1 blocks Kv1.3 in a use-dependent manner and acts by preferentially binding to the C-type inactivated state of the channel. PAP-1 exhibits 23-fold selectivity over Kv1.5 (EC50=45 nM), and further displays 33- to 125-fold selectivity over all other Kv1-family channels. PAP-1 does not exhibit cytotoxic or phototoxic effects.

PAP-1 (5-(4-Phenoxybutoxy)psoralen) is a potent, selective, and orally active Kv1.3 blocker (EC50=2 nM). PAP-1 blocks Kv1.3 in a use-dependent manner and acts by preferentially binding to the C-type inactivated state of the channel. PAP-1 exhibits 23-fold selectivity over Kv1.5 (EC50=45 nM), and further displays 33- to 125-fold selectivity over all other Kv1-family channels. PAP-1 does not exhibit cytotoxic or phototoxic effects.

Specifications

Chemical Name or Material PAP-1
CAS 870653-45-5
Color White
Purity Grade Notes Research
Molecular Formula C21H18O5
Quantity 5 mg
Synonym 5-(4-Phenoxybutoxy)psoralen
Solubility Information DMSO : 50 mg/mL (142.71 mM; Need ultrasonic)
SMILES O=C1C=CC(C(O1)=CC2=C3C=CO2)=C3OCCCCOC4=CC=CC=C4
Molecular Weight (g/mol) 350.36
Formula Weight 350.36
Percent Purity 98.75%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.