Learn More
Epredia™ Papanicolaou EA Stains
Product Code. 10540529 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
Type:
EA-50 Papanicolaou Stain
EA-65 Papanicolaou Stain
Unit Size:
3.79 L
Each
Product Code. 10540529
Brand: Epredia™ 75611
Control the color hues and intensities of your work with Epredia™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining.
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
TRUSTED_SUSTAINABILITY
Specifications
Type EA-65 Papanicolaou Stain
Item Description EA-65; 3.79L (1 Gal.)
Quantity 1 gal.