missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Papilin Antibody (5B2E5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 222.00 - € 568.00
Specifications
| Antigen | Papilin |
|---|---|
| Clone | 5B2E5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18087383
|
Novus Biologicals
NBP2-37320 |
0.1 mL |
€ 568.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643326
|
Novus Biologicals
NBP2-37320-0.025ml |
0.025 mL |
€ 222.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Papilin Monoclonal specifically detects Papilin in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Papilin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 89932 | |
| Purified recombinant fragment of human PAPLN (AA: 766-870) expressed in E. Coli. | |
| Primary | |
| 137.7 kDa |
| 5B2E5 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Human, Mouse | |
| papilin, Papilin, Proteoglycan-Like Sulfated Glycoprotein, PAPLN | |
| PAPLN | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title