missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pappalysin-1/PAPP-A Antibody (10E1cc), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-8346V
This item is not returnable.
View return policy
Description
Pappalysin-1/PAPP-A Monoclonal antibody specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, Immunoprecipitation, Sandwich ELISA
Specifications
| Pappalysin-1/PAPP-A | |
| Monoclonal | |
| DyLight 405 | |
| ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified human PAPP-A/proMBP (heterotetrameric PAPP-A, htPAPP-A) complex. | |
| 0.2 mL | |
| Cancer | |
| 5069 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunoprecipitation, Sandwich ELISA | |
| 10E1cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction