missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pappalysin-1/PAPP-A Antibody (7A6), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73060AF488
This item is not returnable.
View return policy
Description
Pappalysin-1/PAPP-A Monoclonal specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications
| Pappalysin-1/PAPP-A | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PAPPA | |
| purified human PAPP-A/proMBP (heterotetrameric PAPP-A, htPAPP-A) complex. | |
| 0.2 mL | |
| Cancer | |
| 5069 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 7A6 | |
| Western Blot, ELISA, Sandwich ELISA | |
| ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human PAPP-A antigen purified from placental blood (heterotetrameric complex, consisting of PAPP-A subunits and pro-MBP subunits). | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction