missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPSS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14708-1-AP-150UL
This item is not returnable.
View return policy
Description
Three-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) is the sulfate donor cosubstrate for all sulfotransferase (SULT) enzymes (Xu et al., 2000 ). SULTs catalyze the sulfate conjugation of many endogenous and exogenous compounds, including drugs and other xenobiotics. In humans, PAPS is synthesized from adenosine 5-prime triphosphate (ATP) and inorganic sulfate by 2 isoforms, PAPSS1 and PAPSS2.Specifications
| PAPSS1 | |
| Polyclonal | |
| Unconjugated | |
| PAPSS1 | |
| APS kinase, ATP sulfurylase, ATPSK1, PAPS synthase 1, PAPSS, PAPSS 1, PAPSS1, SAT Adenylyl sulfate kinase, SK 1, SK1, Sulfate adenylate transferase, Sulfurylase kinase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23971, 295443, 9061 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43252, Q60967 | |
| PAPSS1 | |
| PAPSS1 Fusion Protein Ag6401 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction