missing translation for 'onlineSavingsMsg'
Learn More

PAPSS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16883484 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16883484 20 μL 20µL
16873484 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16883484 Supplier Proteintech Supplier No. 147081AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Three-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) is the sulfate donor cosubstrate for all sulfotransferase (SULT) enzymes (Xu et al., 2000 ). SULTs catalyze the sulfate conjugation of many endogenous and exogenous compounds, including drugs and other xenobiotics. In humans, PAPS is synthesized from adenosine 5-prime triphosphate (ATP) and inorganic sulfate by 2 isoforms, PAPSS1 and PAPSS2.

Specifications

Antigen PAPSS1
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PAPSS1
Gene Accession No. O43252, Q60967
Gene Alias APS kinase, ATP sulfurylase, ATPSK1, PAPS synthase 1, PAPSS, PAPSS 1, PAPSS1, SAT Adenylyl sulfate kinase, SK 1, SK1, Sulfate adenylate transferase, Sulfurylase kinase 1
Gene Symbols PAPSS1
Host Species Rabbit
Immunogen PAPSS1 Fusion Protein Ag6401
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23971, 295443, 9061
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.