missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPSS2 Monoclonal Antibody (OTI6H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807127S
This item is not returnable.
View return policy
Description
PAPSS2 Monoclonal antibody specifically detects PAPSS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| PAPSS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00797 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5972 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI6H10 | |
| Unconjugated | |
| Ren | |
| 1810018P12Rik; 3'-phosphoadenosine 5'-phosphosulfate synthase 2; 3'-phosphoadenosine-5'-phosphosulfate synthase; 3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2; adenosine 5'-phosphosulfate kinase; Adenosine-5'-phosphosulfate 3'-phosphotransferase; Adenylylsulfate 3'-phosphotransferase; Adenylyl-sulfate kinase; AI159688; APS kinase; ATP sulfurylase/adenosine 5'-phosphosulfate kinase; ATP sulfurylase/APS kinase 2; ATPSK2; AtpsU2; ATP-sulfurylase; BCYM4; Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2; bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2; bm; brachymorphic; PAPS synthase 2; PAPS synthetase 2; PAPS synthethase 2; PAPSS 2; PAPSS2; SAT; SK 2; SK2; Sulfate adenylate transferase; Sulfate adenylyltransferase; sulfurylase kinase 2 | |
| Human recombinant protein fragment corresponding to amino acids 131-406 of human REN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction