missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAPSS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140275
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DTLLDGMALP DGVINMSIPI VLPVSAEDKT R Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 81%.
Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3'-phosphoadenosine 5'-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene.
Specifications
| PAPSS2 | |
| Polyclonal | |
| Unconjugated | |
| PAPSS2 | |
| 1810018P12Rik; 3'-phosphoadenosine 5'-phosphosulfate synthase 2; 3'-phosphoadenosine-5'-phosphosulfate synthase; 3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2; adenosine 5'-phosphosulfate kinase; Adenosine-5'-phosphosulfate 3'-phosphotransferase; Adenylylsulfate 3'-phosphotransferase; Adenylyl-sulfate kinase; AI159688; APS kinase; ATP sulfurylase/adenosine 5'-phosphosulfate kinase; ATP sulfurylase/APS kinase 2; ATPSK2; AtpsU2; ATP-sulfurylase; BCYM4; Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2; bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2; bm; brachymorphic; PAPS synthase 2; PAPS synthetase 2; PAPS synthethase 2; PAPSS 2; PAPSS2; SAT; SK 2; SK2; Sulfate adenylate transferase; Sulfate adenylyltransferase; sulfurylase kinase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9060 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95340 | |
| PAPSS2 | |
| Recombinant protein corresponding to Human PAPSS2. Recombinant protein control fragment (Product #RP-109467). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction