missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR1/Thrombin Receptor Antibody (731115), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3855K
This item is not returnable.
View return policy
Description
PAR1/Thrombin Receptor Monoclonal antibody specifically detects PAR1/Thrombin Receptor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| PAR1/Thrombin Receptor | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 731115 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, TRproteinase-activated receptor 1 | |
| E. coli-derived recombinant human PAR1, Arg27-Thr102, Ser375-Thr425, Accession # P25116 | |
| 0.1 mL | |
| Apoptosis, Cancer, GPCR | |
| 2149 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction