missing translation for 'onlineSavingsMsg'
Få mere at vide

Novus Biologicals™ PAR2 Antibody (Regeneron patent anti-PAR-2), Novus Biologicals™

Artikelnummer. p-200086499
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 mg
100 μg
50 μg
Pakningsstørrelse:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30047216 100 μg 100µg
30044699 1 mg 1mg
30045214 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30047216 Leverandør Novus Biologicals™ Leverandørnr. NBP328886100ug

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Human Monoclonal Antibody

PAR2 Monoclonal antibody specifically detects PAR2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen PAR2
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Regeneron patent anti-PAR-2
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No. P55085
Gene Alias coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1
Host Species Human
Immunogen PAR2
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2150
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Form Purified
Isotype IgG1
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.