Learn More
Paradol, MedChemExpress
Product Code. 16486506
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16486506
Brand: Medchem Express HY14617
MedChemExpress Paradol is a pungent phenolic substance found in ginger and other Zingiberaceae plants. Paradol is an effective inhibitor of tumor promotion in mouse skin carcinogenesis, binds to cyclooxygenase (COX)-2 active site.
Paradol is a pungent phenolic substance found in ginger and other Zingiberaceae plants. Paradol is an effective inhibitor of tumor promotion in mouse skin carcinogenesis, binds to cyclooxygenase (COX)-2 active site.
Specifications
Chemical Name or Material Paradol
CAS 27113-22-0
Color Red
Purity Grade Notes Research
Molecular Formula C17H26O3
Quantity 100 mg
Synonym [6]-Gingerone [6]-Paradol
Solubility Information DMSO : ≥ 140 mg/mL (502.89 mM)
SMILES COC1=C(O)C=CC(CCC(CCCCCCC)=O)=C1
Molecular Weight (g/mol) 278.39
Formula Weight 278.39
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only