Learn More
Paradol, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 6-Paradol |
| CAS | 27113-22-0 |
| Molecular Formula | C17 H26 O3 |
| Quantity | 50 mg |
| Synonym | 6-Gingerone, 1-(4-Hydroxy-3-methylphenol)-3-decanone |
| InChI Formula | InChI=1S/C17H26O3/c1-3-4-5-6-7-8-15(18)11-9-14-10-12-16(19)17(13-14)20-2/h10,12-13,19H,3-9,11H2,1-2H3 |
| SMILES | CCCCCCCC(=O)CCc1ccc(O)c(OC)c1 |
| IUPAC Name | 1-(4-hydroxy-3-methoxy-phenyl)decan-3-one |
| Molecular Weight (g/mol) | 278.39 |
| Formula Weight | 278.1882 |