missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paralemmin 3 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81979V
This item is not returnable.
View return policy
Description
Paralemmin 3 Polyclonal antibody specifically detects Paralemmin 3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Paralemmin 3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 405 | |
| PALM3, paralemmin-3 | |
| Paralemmin3 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human Paralemmin3. The immunogen is located within amino acids 50 - 100 of Paralemmin3. Amino Acid Squence: PEPSEDPTSKDPQSPEGQA | |
| 0.1 mL | |
| Innate Immunity | |
| 342979 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction