missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paralemmin Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97246JF646
This item is not returnable.
View return policy
Description
Paralemmin Polyclonal antibody specifically detects Paralemmin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Paralemmin | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| KIAA0270, paralemmin | |
| Produced in rabbits immunized with E. coli-derived Human Paralemmin fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 5064 | |
| Store at 4°C in the dark. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?