missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Juchheim Laborgeraete™ Parallel Action Clamp
Click to view available options
Height (Metric):
20 mm
30 mm
Unit Size:
Each
Description
- Made of particularly robust malleable cast iron
- With cork insert to protect the clamped glass
- Span up to 130 mm
- Weight: 750 g
Specifications
Specifications
| Material | Tempered Cast Iron |
| Height (Metric) | 20 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction