Learn More
Paramyxovirus SV5 Pk Antibody (SV5-Pk1), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-62264MFV450
Product Code. 18746792
  • € 618.00 / 0.10mL
Description
Paramyxovirus SV5 Pk Monoclonal antibody specifically detects Paramyxovirus SV5 Pk in Mouse, Virus samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Paramyxovirus SV5 Pk
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Protein G purified
RUO
Mouse, Virus
Purified
Flow Cytometry
SV5-Pk1
Flow Cytometry
Simian virus 5; SPV5gp2; V protein
Mice were infected with the paramyxovirus SV5, Simian-Virus 5
0.1 mL
Primary
Store at 4C in the dark.
IgG2a