missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paramyxovirus SV5 Pk Antibody (SV5-Pk1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62264PCP
This item is not returnable.
View return policy
Description
Paramyxovirus SV5 Pk Monoclonal specifically detects Paramyxovirus SV5 Pk in Mouse, Virus samples. It is validated for Flow Cytometry.
Specifications
| Paramyxovirus SV5 Pk | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| This product recognises a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5. NB100-62264 has been used to detect recombinant proteins, some of which include transmembrane and secreted proteins, which have been tagged with this epitope. Usually, a 14 amino acid tag has been added to the recombinant proteins, although a smaller epitope of 9 amino acids (that as a peptide inhibit the binding of the monoclonal antibody to its native protein) has also been successfully used. The 14 amino acid epitope is; gly lys pro (ile pro asn pro leu leu gly leu asp) ser thr. (9 amino acid epitope is in parenthesis) | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| SV5-Pk1 | |
| Flow Cytometry | |
| Simian virus 5; SPV5gp2; V protein | |
| Mice were infected with the paramyxovirus SV5, Simian-Virus 5 | |
| 0.1 mL | |
| Primary | |
| Mouse, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction