missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARC Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77222AF532
This item is not returnable.
View return policy
Description
PARC Polyclonal antibody specifically detects PARC in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry
Specifications
| PARC | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence, Immunocytochemistry | |
| Alexa Fluor 532 | |
| CUL-9, cullin 9, cullin-9, DKFZp686G1042, H7AP1DKFZp686P2024, p53-associated parkin-like cytoplasmic protein, PARCKIAA0708, parkin-like cytoplasmic p53 binding protein, RP3-330M21.2, UbcH7-associated protein 1 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human PARC. The immunogen is located within the last 50 amino acids of PARC. Amino Acid Squence: DNDSFSYDESENLDQET | |
| 0.1 mL | |
| Cancer, DNA Repair, Tumor Suppressors | |
| 23113 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction