missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PARD3/Par3 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
250µg
Specifications
Specifications
| Gene ID (Entrez) | 56288 |
| Species | Mouse |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP2-11253 |
| Gene Alias | FLJ21015, par-3 partitioning defective 3 homolog (C. elegans), PAR3A, PAR3C.elegans) homolog |
| Gene Symbol | PARD3 |
| Quantity | 0.25 mg |
| Research Category | Cell Cycle and Replication |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction