missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARD3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557214
This item is not returnable.
View return policy
Description
Immunogen sequence: ARREGFPLYE DDEGRARPSE YDLLWVPGRG PDGNAHNLRF EGMERQYASL PRGGPADPVD YLPAAPRGLY KERELPYYPG AHPMHPPKGS YPRPTELRVA DLRYPQ Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.
Specifications
| PARD3B | |
| Polyclonal | |
| Unconjugated | |
| Pard3b | |
| 1810008K04Rik; 2010002N16Rik; 2810455B10Rik; Als2cr19; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 19; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein homolog; C. elegans; DN-276O13.2; par-3 family cell polarity regulator beta; par-3 partitioning defective 3 homolog B; PAR3B; PAR3beta; PAR3-beta; PAR3L; PAR3-L protein; Pard3b; Partitioning defective 3 homolog B; partitioning defective 3-like protein; partitioning-defective 3-beta; RP23-20G7.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117583 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TEW8 | |
| Pard3b | |
| Recombinant protein corresponding to Human PARD3B. Recombinant protein control fragment (Product #RP-96042). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction